Skip to Content
Merck
Allylpalladium(II) chloride dimer 98%

Allylpalladium(II) chloride dimer

Allylpalladium(II) chloride dimer 98%
Synonym(s):
[PdCl(C3H5)]2
Empirical Formula (Hill Notation):
C6H10Cl2Pd2
CAS Number:
Molecular Weight:
365.89
EC Number:

Products

Brand
Sustainability
Feature
Formula Weight
Physical Form
Purity
Quality Segment
Markush Class
Markush Group
Reaction Type
Reagent Type
Core
Showing 1-1 of 1
Compare
Product Number
Description
Pricing