Aleuritic acid
Synonym(s):
DL-erythro--9,10,16-Trihydroxypalmitic acid, Aleuritic acid
Empirical Formula (Hill Notation):
C16H32O5
CAS Number:
Molecular Weight:
304.42
Products
Brand
Formula Weight
Manufacturer
Melting Point (°C)
Physical Form
Agency
Format
Quality Segment
Application
Package Size
API Family
Reference Material Type
Special Grade
Showing 1-2 of 2