Skip to Content
Merck
Alcohol Pharmaceutical Secondary Standard; Certified Reference Material

Alcohol

Alcohol Pharmaceutical Secondary Standard; Certified Reference Material
Synonym(s):
Ethanol, Ethyl alcohol
Linear Formula:
CH3CH2OH
CAS Number:
Molecular Weight:
46.07
EC Number:

Products

Brand
Boiling Point (°C)
Formula Weight
Manufacturer
Agency
Format
Quality Segment
Application
Markush Class
Markush Group
Package Size
API Family
Reference Material Type
Special Grade
Technique