Alantolactone
Synonym(s):
(+)-Alantolactone., 3aR,5S,8aR,9aR)-3a,5,6,7,8,8a,9,9a-Octahydro-5,8a-dimethyl-3-methylene-naphtho[2,3-b]furan-2(3H)-one, Alant camphor, Elecampane camphor, Eupatal, Inula camphor, NSC 333843, NSC 93131
Empirical Formula (Hill Notation):
C15H20O2
Products
Product Category
Brand
Color
Formula Weight
Manufacturer
Physical Form
Product Line
Purity
Format
Research Area
Application
Reference Material Type
Special Grade
Showing 1-2 of 2
Compare
Product Number
Description
Pricing