Skip to Content
Merck
Acetoin May exist as crystalline dimer

Acetoin

Acetoin May exist as crystalline dimer
Synonym(s):
3-Hydroxy-2-butanone, Acetylmethylcarbinol
Linear Formula:
CH3COCH(OH)CH3
CAS Number:
Molecular Weight:
88.11
EC Number:

Products

Product Category
Brand
Sustainability
Biological Source
Boiling Point (°C)
Feature
Formula Weight
Melting Point (°C)
Physical Form
Purity
Agency
Format
Quality Segment
Application
Markush Class
Markush Group
Organoleptic Class
Reference Material Type
Special Grade
Technique
Showing 1-4 of 4
Compare
Product Number
Description
Pricing