Acarbose for peak identification
Synonym(s):
Acarbose, 4",6"-Dideoxy-4"-([1S]-[1,4,6/5]-4,5,6-trihydroxy-3-hydroxymethyl-2-yclohexenylamino)-maltotriose
Empirical Formula (Hill Notation):
C25H43NO18
CAS Number:
Molecular Weight:
645.60
Products
Formula Weight
Gene Alias
Manufacturer
Format
Application
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing