Abiraterone acetate
Synonym(s):
17-(Pyridin-3-yl)androsta-5,16-dien-3β-yl acetate, Abiraterone acetate
Empirical Formula (Hill Notation):
C26H33NO2
CAS Number:
Molecular Weight:
391.55
Products
Product Category
Brand
Color
Formula Weight
Gene Alias
Manufacturer
Physical Form
Purity
Agency
Format
Research Area
Quality Segment
Application
Package Size
API Family
Reference Material Type
Special Grade
Showing 1-3 of 3