Skip to Content
Merck
[8]-Gingerol analytical standard

[8]-Gingerol

[8]-Gingerol analytical standard
Synonym(s):
(S)-5-Hydroxy-1-(4-hydroxy-3-methoxyphenyl)-3-dodecanone, 8-Gingerol, [8]-Gingerol
Empirical Formula (Hill Notation):
C19H30O4
CAS Number:
Molecular Weight:
322.44

Products

Product Category
Brand
Color
Formula Weight
Manufacturer
Physical Form
Product Line
Purity
Format
Research Area
Quality Segment
Application
Reference Material Type
Special Grade
Technique
Showing 1-3 of 3
Compare
Product Number
Description
Pricing