![8-Acetylharpagide phyproof® Reference Substance](/deepweb/assets/sigmaaldrich/product/structures/844/556/df2a4a68-4c9a-4a8b-ae30-f24bec03a558/640/df2a4a68-4c9a-4a8b-ae30-f24bec03a558.png)
8-Acetylharpagide
![8-Acetylharpagide phyproof® Reference Substance](/deepweb/assets/sigmaaldrich/product/structures/844/556/df2a4a68-4c9a-4a8b-ae30-f24bec03a558/640/df2a4a68-4c9a-4a8b-ae30-f24bec03a558.png)
Synonym(s):
Harpagide 8-acetate
Empirical Formula (Hill Notation):
C17H26O11
CAS Number:
Molecular Weight:
406.38
Products
Product Category
Brand
Formula Weight
Manufacturer
Physical Form
Product Line
Purity
Research Area
Application
Reference Material Type
Special Grade
Showing 1-2 of 2
Compare
Product Number
Description
Pricing