![7-O-Acetylpaclitaxel certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland](/deepweb/assets/sigmaaldrich/product/structures/202/109/e0cbe85f-1d5f-4df3-a076-2b0ef6a510cb/640/e0cbe85f-1d5f-4df3-a076-2b0ef6a510cb.png)
7-O-Acetylpaclitaxel
![7-O-Acetylpaclitaxel certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland](/deepweb/assets/sigmaaldrich/product/structures/202/109/e0cbe85f-1d5f-4df3-a076-2b0ef6a510cb/640/e0cbe85f-1d5f-4df3-a076-2b0ef6a510cb.png)
Synonym(s):
7-Acetyltaxol
Empirical Formula (Hill Notation):
C49H53NO15
CAS Number:
Molecular Weight:
895.94
Products
Brand
Formula Weight
Manufacturer
Physical Form
Product Line
Quality Segment
Application
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing