Skip to Content
Merck
7-Hydroxyflavanone 98%

7-Hydroxyflavanone

7-Hydroxyflavanone 98%
Synonym(s):
7-Hydroxy-2-phenylchroman-4-one
Empirical Formula (Hill Notation):
C15H12O3
CAS Number:
Molecular Weight:
240.25

Products

Product Category
Brand
Formula Weight
Gene Alias
Melting Point (°C)
Physical Form
Purity
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing