Skip to Content
Merck
6-Maleimidohexanoic acid ≥98.0% (HPLC)

6-Maleimidohexanoic acid

6-Maleimidohexanoic acid ≥98.0% (HPLC)
Synonym(s):
6-Maleimidocaproic acid, N-Maleoyl-6-aminocaproic acid, N-(5-Carboxypentyl)maleimide
Empirical Formula (Hill Notation):
C10H13NO4
CAS Number:
Molecular Weight:
211.21

Products

Product Category
Brand
Formula Weight
Melting Point (°C)
Physical Form
Purity
Quality Segment
Markush Class
Markush Group
Showing 1-2 of 2
Compare
Product Number
Description
Pricing