
6-Hydroxyflavanone

Empirical Formula (Hill Notation):
C15H12O3
CAS Number:
Molecular Weight:
240.25
Products
Product Category
Brand
Formula Weight
Gene Alias
Melting Point (°C)
Physical Form
Purity
Quality Segment
Functional Group
Markush Class
Markush Group
Showing 1-1 of 1