![6-Hydroxychlorzoxazone ≥98% (HPLC)](/deepweb/assets/sigmaaldrich/product/structures/248/102/9d419a16-d859-444a-bcdd-73489420e667/640/9d419a16-d859-444a-bcdd-73489420e667.png)
6-Hydroxychlorzoxazone
![6-Hydroxychlorzoxazone ≥98% (HPLC)](/deepweb/assets/sigmaaldrich/product/structures/248/102/9d419a16-d859-444a-bcdd-73489420e667/640/9d419a16-d859-444a-bcdd-73489420e667.png)
Synonym(s):
5-Chloro-6-hydroxy-2(3H)-benzoxazolone, 5-Chloro-6-hydroxybenzoxazone
Empirical Formula (Hill Notation):
C7H4ClNO3
CAS Number:
Molecular Weight:
185.56
Products
Product Category
Brand
Color
Formula Weight
Physical Form
Purity
Research Area
Quality Segment
Showing 1-1 of 1
Compare
Product Number
Description
Pricing