Skip to Content
Merck
5-Methylthiazole 97%

5-Methylthiazole

5-Methylthiazole 97%
Empirical Formula (Hill Notation):
C4H5NS
CAS Number:
Molecular Weight:
99.15

Products

Product Category
Brand
Boiling Point (°C)
Formula Weight
Physical Form
Purity
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing