
5-(Difluoromethoxy)-2-mercapto-1H-benzimidazole

Empirical Formula (Hill Notation):
C8H6F2N2OS
CAS Number:
Molecular Weight:
216.21
Products
Product Category
Formula Weight
Melting Point (°C)
Purity
Functional Group
Markush Class
Markush Group
Showing 1-1 of 1