Skip to Content
Merck
(5-Bromopentyl)trimethylammonium bromide 97%

(5-Bromopentyl)trimethylammonium bromide

(5-Bromopentyl)trimethylammonium bromide 97%
Linear Formula:
Br(CH2)5N(CH3)3Br
CAS Number:
Molecular Weight:
289.05

Products

Brand
Formula Weight
Melting Point (°C)
Purity
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing