Skip to Content
Merck
5,6-Benzo-4,7,13,16,21,24-hexaoxa-1,10-diazabicyclo[8.8.8]hexacos-5-ene solution 50 wt. % in toluene

5,6-Benzo-4,7,13,16,21,24-hexaoxa-1,10-diazabicyclo[8.8.8]hexacos-5-ene solution

5,6-Benzo-4,7,13,16,21,24-hexaoxa-1,10-diazabicyclo[8.8.8]hexacos-5-ene solution 50 wt. % in toluene
Synonym(s):
Kryptand 222B
Empirical Formula (Hill Notation):
C22H36N2O6
CAS Number:
Molecular Weight:
424.53
EC Number:

Products

Brand
Formula Weight
Physical Form
Markush Class
Markush Group
Showing 1-1 of 1