Skip to Content
AU
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Water Purification
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
Get Site Smart
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
5,5′-Dithiobis(2-nitrobenzoic acid)
Synonym(s):
3-Carboxy-4-nitrophenyl disulfide, 6,6′-Dinitro-3,3′-dithiodibenzoic acid, Bis(3-carboxy-4-nitrophenyl) disulfide, DTNB, Ellman’s Reagent, 5,5′-Dithio-
bis
-(2-nitrobenzoic Acid)
Linear Formula:
[-SC
6
H
3
(NO
2
)CO
2
H]
2
CAS Number:
69-78-3
Molecular Weight:
396.35
EC Number:
200-714-4
Products
Product Category
building blocks (1)
Brand
Sigma-Aldrich (2)
Color
Formula Weight
Melting Point (°C)
Physical Form
Product Line
Purity
Quality Segment
Markush Class
Markush Group
Filter Products
Showing 1-4 of 4
Compare
Product Number
Description
Pricing
D8130
5,5′-Dithiobis(2-nitrobenzoic acid),
≥98%, BioReagent, suitable for determination of sulfhydryl groups
Expand
View Pricing
D218200
5,5′-Dithiobis(2-nitrobenzoic acid),
ReagentPlus
®
, 99%
Expand
View Pricing
322123
5,5′-Dithiobis(2-nitrobenzoic acid),
Sulfhydryl reagent used to characterize reactive SH groups.
Expand
View Pricing
V900454
5,5′-Dithiobis(2-nitrobenzoic acid),
Vetec
™
, reagent grade, 98%
Expand
View Pricing