Skip to Content
Merck
4-(Trimethylsilyl)morpholine 97%

4-(Trimethylsilyl)morpholine

4-(Trimethylsilyl)morpholine 97%
Empirical Formula (Hill Notation):
C7H17NOSi
CAS Number:
Molecular Weight:
159.30

Products

Product Category
Brand
Boiling Point (°C)
Formula Weight
Physical Form
Purity
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing