4-tert-Butylphenylmagnesium bromide solution
Linear Formula:
(CH3)3CC6H4MgBr
CAS Number:
Molecular Weight:
237.42
Products
Brand
Boiling Point (°C)
Formula Weight
Physical Form
Markush Class
Markush Group
Reaction Type
Showing 1-2 of 2
Compare
Product Number
Description
Pricing