Skip to Content
Merck
(4R,5R)-2,2-Dimethyl-α,α,α′,α′-tetra(2-naphthyl)dioxolane-4,5-dimethanol ≥99.0% (sum of enantiomers, HPLC)

(4R,5R)-2,2-Dimethyl-α,α,α′,α′-tetra(2-naphthyl)dioxolane-4,5-dimethanol

(4R,5R)-2,2-Dimethyl-α,α,α′,α′-tetra(2-naphthyl)dioxolane-4,5-dimethanol ≥99.0% (sum of enantiomers, HPLC)
Synonym(s):
(−)-2,3-O-Isopropylidene-1,1,4,4-tetra(2-naphthyl)-L-threitol, (−)-DINOL
Empirical Formula (Hill Notation):
C47H38O4
CAS Number:
Molecular Weight:
666.80

Products

Brand
Formula Weight
Melting Point (°C)
Purity
Markush Class
Markush Group
Optical Isomer
Special Grade