![4-Nitroacetanilide 98%](/deepweb/assets/sigmaaldrich/product/structures/218/454/823b6469-3c42-4183-a647-bfe3f0c1513c/640/823b6469-3c42-4183-a647-bfe3f0c1513c.png)
4-Nitroacetanilide
![4-Nitroacetanilide 98%](/deepweb/assets/sigmaaldrich/product/structures/218/454/823b6469-3c42-4183-a647-bfe3f0c1513c/640/823b6469-3c42-4183-a647-bfe3f0c1513c.png)
Synonym(s):
4′-Nitroacetanilide, Acetic acid 4-nitroanilide
Linear Formula:
CH3CONHC6H4-4-(NO2)
CAS Number:
Molecular Weight:
180.16
Products
Product Category
Brand
Formula Weight
Melting Point (°C)
Purity
Quality Segment
Functional Group
Markush Class
Markush Group
Showing 1-2 of 2
Compare
Product Number
Description
Pricing