Skip to Content
Merck
4-Methyl-3,5-dinitrobenzoic acid 98%

4-Methyl-3,5-dinitrobenzoic acid

4-Methyl-3,5-dinitrobenzoic acid 98%
Synonym(s):
3,5-Dinitro-4-methylbenzoic acid, 3,5-Dinitro-p-toluic acid
Linear Formula:
(O2N)2C6H2(CH3)CO2H
CAS Number:
Molecular Weight:
226.14
EC Number:

Products

Product Category
Brand
Formula Weight
Melting Point (°C)
Physical Form
Purity
Quality Segment
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing