4-Hydroxyisophthalic acid
Synonym(s):
4-Hydroxy 1,3-benzenedicarboxylic acid
Linear Formula:
HOC6H3-1,3-(CO2H)2
CAS Number:
Molecular Weight:
182.13
Products
Brand
Formula Weight
Manufacturer
Physical Form
Product Line
Purity
Markush Class
Markush Group
Reference Material Type
Special Grade
Showing 1-2 of 2
Compare
Product Number
Description
Pricing