Skip to Content
Merck
4-Chlorostyrene 97%, contains 500 ppm 4-tert-butylcatechol as inhibitor

4-Chlorostyrene

4-Chlorostyrene 97%, contains 500 ppm 4-tert-butylcatechol as inhibitor
Synonym(s):
4-Chlorostyrene
Linear Formula:
H2C=CHC6H4Cl
CAS Number:
Molecular Weight:
138.59

Products

Brand
Boiling Point (°C)
Formula Weight
Physical Form
Purity
Quality Segment
Markush Class
Markush Group
Monomer Type