4-Aminobenzoylglutamic acid
Synonym(s):
N-(4-Aminobenzoyl)-L-glutamic acid
Empirical Formula (Hill Notation):
C12H14N2O5
CAS Number:
Molecular Weight:
266.25
Products
Formula Weight
Manufacturer
Format
Application
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing