4-Acetylbenzoic acid
Synonym(s):
Acetophenone-4-carboxylic acid, 4-Acetylbenzoic acid
Linear Formula:
CH3COC6H4CO2H
CAS Number:
Molecular Weight:
164.16
Products
Product Category
Brand
Formula Weight
Melting Point (°C)
Physical Form
Purity
Quality Segment
Application
Markush Class
Markush Group
Package Size
API Family
Reference Material Type
Special Grade
Showing 1-2 of 2
Compare
Product Number
Description
Pricing