Skip to Content
Merck
4-Acetoxy-2,5-dimethyl-3(2H)furanone ≥95%, FG

4-Acetoxy-2,5-dimethyl-3(2H)furanone

4-Acetoxy-2,5-dimethyl-3(2H)furanone ≥95%, FG
Synonym(s):
Strawberry acetate
Empirical Formula (Hill Notation):
C8H10O4
CAS Number:
Molecular Weight:
170.16

Products

Product Category
Brand
Biological Source
Boiling Point (°C)
Formula Weight
Purity
Agency
Quality Segment
Application
Organoleptic Class
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing