Skip to Content
Merck
4-Acetamido-TEMPO, free radical 97%

4-Acetamido-TEMPO, free radical

4-Acetamido-TEMPO, free radical 97%
Synonym(s):
4-Acetamido-2,2,6,6-tetramethylpiperidine 1-oxyl
Empirical Formula (Hill Notation):
C11H21N2O2
CAS Number:
Molecular Weight:
213.30
EC Number:

Products

Brand
Formula Weight
Melting Point (Ā°C)
Physical Form
Purity
Showing 1-1 of 1
Compare
Product Number
Description
Pricing