4-[2-[(1,1-Dimethylethyl)amino]ethyl]-2-methylphenol
Synonym(s):
4-[2-(tert-Butylamino)ethyl]-2-methylphenol hydrochloride, 4-[2-[(1,1-Dimethylethyl)amino]ethyl]-2-methylphenol hydrochloride
Empirical Formula (Hill Notation):
C13H21NO · HCl
CAS Number:
Molecular Weight:
243.77
Products
Brand
Formula Weight
Physical Form
Quality Segment
Application
Package Size
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing