Skip to Content
Merck
3-Methyl-2(5H)-furanone technical grade, 90%

3-Methyl-2(5H)-furanone

3-Methyl-2(5H)-furanone technical grade, 90%
Empirical Formula (Hill Notation):
C5H6O2
CAS Number:
Molecular Weight:
98.10

Products

Product Category
Brand
Boiling Point (°C)
Formula Weight
Physical Form
Purity
Markush Class
Markush Group
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing