Skip to Content
AU
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Water Purification
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
Get Site Smart
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
3-Methoxytyramine hydrochloride
Synonym(s):
3-MT, 3-Methoxy-4-hydroxyphenethylamine hydrochloride, 4-(2-Aminoethyl)-2-methoxyphenol hydrochloride
Linear Formula:
CH
3
OC
6
H
3
-4-(OH)CH
2
CH
2
NH
2
·HCl
CAS Number:
1477-68-5
Molecular Weight:
203.67
EC Number:
216-035-1
Products
Product Category
bioactive small molecules (1)
building blocks (1)
metabolites (1)
Brand
Sigma-Aldrich (1)
Supelco (1)
Color
Formula Weight
Gene Alias
Melting Point (°C)
Physical Form
Purity
Format
Research Area
Quality Segment
Functional Group
Application
Markush Class
Markush Group
Special Grade
analytical standard (1)
Technique
Filter Products
Showing 1-3 of 3
Compare
Product Number
Description
Pricing
65390
3-Methoxytyramine hydrochloride,
99% (AT)
Expand
View Pricing
M4251
3-Methoxytyramine hydrochloride,
≥95.5%, crystalline
Expand
View Pricing
75024
3-Methoxytyramine hydrochloride,
analytical standard
Expand
View Pricing