Skip to Content
Merck
3-Ethyl 5-methyl 2-(2-aminoethoxymethyl)-4-(2-chlorophenyl)-6-methylpyridine-3,5-dicarboxylate oxalate salt certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland

3-Ethyl 5-methyl 2-(2-aminoethoxymethyl)-4-(2-chlorophenyl)-6-methylpyridine-3,5-dicarboxylate oxalate salt

3-Ethyl 5-methyl 2-(2-aminoethoxymethyl)-4-(2-chlorophenyl)-6-methylpyridine-3,5-dicarboxylate oxalate salt certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Empirical Formula (Hill Notation):
C20H23ClN2O5 · C2H2O4
CAS Number:
Molecular Weight:
496.89

Products

Brand
Formula Weight
Manufacturer
Product Line
Format
Quality Segment
Application
Special Grade