Skip to Content
Merck
3-epi-25-Hydroxyvitamin D3 solution 100 μg/mL in ethanol, 98% (CP)

3-epi-25-Hydroxyvitamin D3 solution

3-epi-25-Hydroxyvitamin D3 solution 100 μg/mL in ethanol, 98% (CP)
Empirical Formula (Hill Notation):
C27H44O2
CAS Number:
Molecular Weight:
400.64
EC Number:

Products

Product Category
Brand
Color
Formula Weight
Physical Form
Purity
Application
Markush Class
Markush Group
Technique