
3-Chloro-4-fluorophenylmagnesium bromide solution

Empirical Formula (Hill Notation):
C6H3BrClFMg
CAS Number:
Molecular Weight:
233.75
Products
Brand
Boiling Point (°C)
Formula Weight
Functional Group
Markush Class
Markush Group
Reaction Type
Showing 1-1 of 1
Compare
Product Number
Description
Pricing