Skip to Content
Merck
3-Chloro-2,2-dimethyl-1-propanol 99%

3-Chloro-2,2-dimethyl-1-propanol

3-Chloro-2,2-dimethyl-1-propanol 99%
Synonym(s):
2-Chloromethyl-2-methyl-1-propanol
Linear Formula:
ClCH2C(CH3)2CH2OH
CAS Number:
Molecular Weight:
122.59
EC Number:

Products

Product Category
Brand
Boiling Point (°C)
Formula Weight
Melting Point (°C)
Purity
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing