3-[Bis(2-hydroxyethyl)amino]propyl-triethoxysilane solution
Synonym(s):
N-(3-Triethoxysilylpropyl)diethanolamine
Empirical Formula (Hill Notation):
C13H31NO5Si
CAS Number:
Molecular Weight:
309.47
Products
Product Category
Brand
Formula Weight
Physical Form
Markush Class
Markush Group
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing