Skip to Content
Merck
3,5-Bis(trifluoromethyl)phenylacetic acid 98%

3,5-Bis(trifluoromethyl)phenylacetic acid

3,5-Bis(trifluoromethyl)phenylacetic acid 98%
Linear Formula:
(CF3)2C6H3CH2CO2H
CAS Number:
Molecular Weight:
272.14
EC Number:

Products

Product Category
Brand
Formula Weight
Melting Point (°C)
Physical Form
Purity
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing