Skip to Content
Merck
3,4-Diaminobenzophenone 97%

3,4-Diaminobenzophenone

3,4-Diaminobenzophenone 97%
Synonym(s):
4-Benzoyl-o-phenylenediamine
Linear Formula:
C6H5COC6H3(NH2)2
CAS Number:
Molecular Weight:
212.25
EC Number:

Products

Brand
Formula Weight
Melting Point (°C)
Purity
Markush Class
Markush Group
Monomer Type
Showing 1-1 of 1
Compare
Product Number
Description
Pricing