Skip to Content
Merck
3-(4-Chloro-phenyl)-1H-pyrazole-4-carbaldehyde AldrichCPR

3-(4-Chloro-phenyl)-1H-pyrazole-4-carbaldehyde

3-(4-Chloro-phenyl)-1H-pyrazole-4-carbaldehyde AldrichCPR
Empirical Formula (Hill Notation):
C10H7ClN2O
Molecular Weight:
206.63

Products

Product Category
Brand
Formula Weight
Physical Form
Markush Class
Markush Group
Showing 1-2 of 2