Skip to Content
Merck
3,3,4,4,5,5,6,6,7,7,8,8,8-Tridecafluorooctyl acrylate contains inhibitor, 97%

3,3,4,4,5,5,6,6,7,7,8,8,8-Tridecafluorooctyl acrylate

3,3,4,4,5,5,6,6,7,7,8,8,8-Tridecafluorooctyl acrylate contains inhibitor, 97%
Synonym(s):
1H,1H,2H,2H-perfluorooctyl acrylate, TFOA
Linear Formula:
H2C=CHCO2CH2CH2(CF2)5CF3
CAS Number:
Molecular Weight:
418.15
EC Number:

Products

Brand
Boiling Point (°C)
Formula Weight
Purity
Markush Class
Markush Group
Monomer Type
Showing 1-1 of 1
Compare
Product Number
Description
Pricing