Skip to Content
Merck
(2RS)-2-[4-(2-Methylpropyl)phenyl]propan-1-ol certified reference material, pharmaceutical secondary standard

(2RS)-2-[4-(2-Methylpropyl)phenyl]propan-1-ol

(2RS)-2-[4-(2-Methylpropyl)phenyl]propan-1-ol certified reference material, pharmaceutical secondary standard
Synonym(s):
β-Methyl-4-(2-methylpropyl)benzeneethanol, 2-(4-Isobutylphenyl)propanol, 2-(4-Isobutylphenyl)propyl alcohol, Ibuprofen alcohol
Empirical Formula (Hill Notation):
C13H20O
CAS Number:
Molecular Weight:
192.30

Products

Brand
Formula Weight
Quality Segment
Application
Special Grade