2-Naphthylboronic acid
Synonym(s):
2-Naphthaleneboronic acid
Linear Formula:
C10H7B(OH)2
CAS Number:
Molecular Weight:
171.99
Products
Brand
Formula Weight
Melting Point (°C)
Purity
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing