2-Chloroethyl vinyl ether
Synonym(s):
2-Chloroethyl vinyl ether
Linear Formula:
ClCH2CH2OCH=CH2
CAS Number:
Molecular Weight:
106.55
Products
Brand
Boiling Point (°C)
Formula Weight
Physical Form
Purity
Quality Segment
Application
Markush Class
Markush Group
Package Size
Monomer Type
Special Grade
Showing 1-2 of 2