Skip to Content
Merck
2-Chloro-5-nitrobenzoic acid Pharmaceutical Secondary Standard; Certified Reference Material

2-Chloro-5-nitrobenzoic acid

2-Chloro-5-nitrobenzoic acid Pharmaceutical Secondary Standard; Certified Reference Material
Synonym(s):
2-Chloro-5-nitrobenzoic acid
Linear Formula:
ClC6H3(NO2)CO2H
CAS Number:
Molecular Weight:
201.56
EC Number:

Products

Product Category
Brand
Formula Weight
Melting Point (°C)
Physical Form
Purity
Format
Quality Segment
Application
Markush Class
Markush Group
Package Size
API Family
Reference Material Type
Special Grade
Technique