Skip to Content
Merck
2′,7′-Bis(2-carboxyethyl)-5(6)-carboxyfluorescein ≥90% (HPLC)

2′,7′-Bis(2-carboxyethyl)-5(6)-carboxyfluorescein

2′,7′-Bis(2-carboxyethyl)-5(6)-carboxyfluorescein ≥90% (HPLC)
Synonym(s):
BCECF
Empirical Formula (Hill Notation):
C27H20O11
CAS Number:
Molecular Weight:
520.44

Products

Product Category
Brand
Formula Weight
Physical Form
Purity
Quality Segment
Showing 1-1 of 1
Compare
Product Number
Description
Pricing