Skip to Content
Merck
25-Hydroxyvitamin D2 solution 5 μg/mL in ethanol, 98% (CP)

25-Hydroxyvitamin D2 solution

25-Hydroxyvitamin D2 solution 5 μg/mL in ethanol, 98% (CP)
Synonym(s):
25-Hydroxycalciferol solution, 25-Hydroxyergocalciferol solution
Empirical Formula (Hill Notation):
C28H44O2
Molecular Weight:
412.65
EC Number:

Products

Product Category
Brand
Color
Formula Weight
Physical Form
Purity
Quality Segment
Application
Markush Class
Markush Group
Technique
Showing 1-3 of 3
Compare
Product Number
Description
Pricing