Skip to Content
Merck
2,4-Difluorophenyl isocyanate 99%

2,4-Difluorophenyl isocyanate

2,4-Difluorophenyl isocyanate 99%
Linear Formula:
F2C6H3NCO
CAS Number:
Molecular Weight:
155.10

Products

Product Category
Brand
Boiling Point (°C)
Formula Weight
Physical Form
Purity
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing