Skip to Content
Merck
2,4,6-Trimethylphenylboronic acid ≥95%

2,4,6-Trimethylphenylboronic acid

2,4,6-Trimethylphenylboronic acid ≥95%
Synonym(s):
2,4,6-Trimethylbenzeneboronic acid, Mesitylene-2-boronic acid, NSC 157832
Linear Formula:
C6H2(CH3)3B(OH)2
CAS Number:
Molecular Weight:
164.01

Products

Formula Weight
Melting Point (°C)
Physical Form
Purity
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing